Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Benzoylpropionic acid | 2051-95-8 | 178.18 | 25 G
SDP

Catalog No. 5000450094
Encompass_Preferred

3-Benzoylpropionic acid is an intermediate used in organic synthesis for the preparation of other organic compounds. It is supplied as a white to light yellow solid, suitable for various laboratory applications.

  • Purity of 99.97%
  • CAS number: 2051-95-8
  • Molecular formula: C10H10O3
  • Molecular weight: 178.19 g/mol
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 200 mg/mL
  • For research use only

Catalog No. 50-004-50094 Supplier Medchemexpress LLC Supplier No. 25GHYW001943
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3-Benzoylpropionic acid is an intermediate used in organic synthesis for the preparation of other organic compounds. It is supplied as a white to light yellow solid, suitable for various laboratory applications.

  • Purity of 99.97%
  • CAS number: 2051-95-8
  • Molecular formula: C10H10O3
  • Molecular weight: 178.19 g/mol
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 200 mg/mL
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.