Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Benzyl-6-bromo-1H-quinolin-2-one | 924633-09-0 | 98.3% | 314.18 | 100 MG
SDP

Catalog No. 5000428843
Encompass_Preferred

3-Benzyl-6-bromo-1H-quinolin-2-one is a drug intermediate used for the synthesis of various active compounds. It is supplied as a white to off-white solid with a purity of 98.33%.

  • Purity of 98.33%
  • Molecular weight of 314.18
  • CAS number 924633-09-0
  • Solid appearance
  • White to off-white color
  • Stable as powder for 3 years at -20°C or 2 years at 4°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C

Catalog No. 50-004-28843 Supplier Medchemexpress LLC Supplier No. HYI1084100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Benzyl-6-bromo-1H-quinolin-2-one is a drug intermediate used for the synthesis of various active compounds. It is supplied as a white to off-white solid with a purity of 98.33%.

  • Purity of 98.33%
  • Molecular weight of 314.18
  • CAS number 924633-09-0
  • Solid appearance
  • White to off-white color
  • Stable as powder for 3 years at -20°C or 2 years at 4°C
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.