Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(benzyl(isopropyl)amino)-1-(naphthalen-2-yl)propan-1-one hydrochloride | 1021868-92-7 | MFCD04974534 | >98.0% | 367.91 g/mol | C23H26ClNO | 10MM 1ML
SDP

Catalog No. 5000235327
Encompass_Preferred

ZM39923 hydrochloride is a small-molecule JAK3 inhibitor used in research. It displays JAK3 potency (pIC50 = 7.1) and also inhibits tissue transglutaminase (TGM2) with an IC50 of 10 nM; it is supplied as a 10 mM solution in DMSO and in solid quantities.

  • Small-molecule JAK3 inhibitor (pIC50 = 7.1).
  • Potent TGM2 inhibition (IC50 = 10 nM).
  • Available as 10 mM solution in DMSO and in solid forms.
  • Molecular formula C23H26ClNO.
  • Molecular weight 367.91 g/mol.
  • Purity >98.0% by HPLC.
  • For research use only.

Catalog No. 50-002-35327 Supplier Medchemexpress LLC Supplier No. HY1258910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZM39923 hydrochloride is a small-molecule JAK3 inhibitor used in research. It displays JAK3 potency (pIC50 = 7.1) and also inhibits tissue transglutaminase (TGM2) with an IC50 of 10 nM; it is supplied as a 10 mM solution in DMSO and in solid quantities.

  • Small-molecule JAK3 inhibitor (pIC50 = 7.1).
  • Potent TGM2 inhibition (IC50 = 10 nM).
  • Available as 10 mM solution in DMSO and in solid forms.
  • Molecular formula C23H26ClNO.
  • Molecular weight 367.91 g/mol.
  • Purity >98.0% by HPLC.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.