Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Benzyloxy)-1-propanol 98.0+%, TCI America™
SDP

Catalog No. B32775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32775G 5 g
B327725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32775G Supplier TCI America Supplier No. B32775G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4799-68-2
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00029659
InChI Key FUCYABRIJPUVAT-UHFFFAOYSA-N
Synonym 3-benzyloxy-1-propanol, 3-benzyloxy propan-1-ol, 3-benzyloxypropanol, 1-propanol, 3-phenylmethoxy, 3-benzyloxy-propan-1-ol, 1-propanol, 3-benzyloxy, 3-benzyloxypropan-1-ol, 3-benzyloxy-propane-1-ol, 3-benzyl-oxy propan-1-ol
PubChem CID 347971
IUPAC Name 3-(benzyloxy)propan-1-ol
SMILES OCCCOCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 120°C
Quantity 5 g
Formula Weight 166.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Benzyloxy)-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.