Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Benzyloxy-4-methoxybenzaldehyde 97.0+%, TCI America™
SDP

Catalog No. B288525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B288525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B288525G Supplier TCI America Supplier No. B288525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6346-05-0
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00003386
InChI Key VQVQZFHUXRSRBZ-UHFFFAOYSA-N
Synonym 3-benzyloxy-4-methoxybenzaldehyde, o-benzylisovanillin, benzaldehyde, 4-methoxy-3-phenylmethoxy, benzyl iso-vanillin, 4-methoxy-3-phenylmethoxy benzaldehyde, 3-benzyloxy-4-methoxy-benzaldehyde, benzylisovanillin, pubchem13136, acmc-1b6ff
PubChem CID 80671
IUPAC Name 3-(benzyloxy)-4-methoxybenzaldehyde
SMILES COC1=CC=C(C=O)C=C1OCC1=CC=CC=C1

Specifications

Melting Point 63°C
Color Yellow
Boiling Point 120°C
Quantity 25 g
Formula Weight 242.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzyloxy-4-methoxybenzaldehyde
EINECSNumber (3)-2922
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.