Learn More
Aobchem 3-(Benzyloxy)pyridine | ≥95% | CAS 76509-17-6 | C12H11NO | 1g
Catalog No. 5000906068
Encompass_Preferred
3-(Benzyloxy)pyridine | ≥95% | CAS 76509-17-6 | C12H11NO | 1g
Catalog No. 50-009-06068 Supplier Aobchem Supplier No. A1001511G
3-(Benzyloxy)pyridine | ≥95% | CAS 76509-17-6 | C12H11NO | 1g