Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Benzyloxybenzeneboronic acid, 98+%, Thermo Scientific™

Catalog No. AAL1747403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1747403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1747403 Supplier Thermo Scientific Chemicals Supplier No. L1747403

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156682-54-1
Molecular Formula C13H13BO3
Molecular Weight (g/mol) 228.05
MDL Number MFCD02093063
InChI Key WIJNYNBSPQMJGO-UHFFFAOYSA-N
Synonym 3-benzyloxyphenylboronic acid, 3-benzyloxybenzeneboronic acid, 3-benzyloxy phenyl boronic acid, 3-benzyloxy phenylboronic acid, m-benzyloxyphenylboronic acid, 3-benzyloxyphenyl boronic acid, 3-boronophenoxy methyl benzene, 3-benzyl-oxy phenyl boranediol, boronic acid, 3-phenylmethoxy phenyl
PubChem CID 2734313
IUPAC Name (3-phenylmethoxyphenyl)boronic acid
SMILES OB(O)C1=CC(OCC2=CC=CC=C2)=CC=C1

Specifications

Melting Point 122°C to 124°C
Quantity 1 g
Beilstein 6808244
Formula Weight 228.06
Percent Purity ≥98%
Chemical Name or Material 3-Benzyloxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.