Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Benzylpyridine 98.0+%, TCI America™
SDP

Catalog No. B155310G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B155310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B155310G Supplier TCI America Supplier No. B155310G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 620-95-1
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.227
MDL Number MFCD00006408
InChI Key UUCLVSDUMKMBSM-UHFFFAOYSA-N
PubChem CID 12112
IUPAC Name 3-benzylpyridine
SMILES C1=CC=C(C=C1)CC2=CN=CC=C2

Specifications

Color White-Yellow
Quantity 10 g
Formula Weight 169.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Benzylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.