Learn More
Medchemexpress LLC 3-(benzylthio)-5-(phenoxymethyl)-4-phenyl-4H-1,2,4-triazole | 129544-85-0 | 99.9% | C22H19N3OS | 10MG
SDP
Catalog No. 5000258837
Encompass_Preferred
MIND4-19 is a small-molecule SIRT2 inhibitor (IC50 7.0 μM) used as a research tool in studies of neurodegeneration, including Huntington's disease. The compound is a solid, white to off-white material with formula C22H19N3OS and a molecular weight of 373.47 g·mol⁻¹; it is supplied with high purity and storage recommendations to protect activity.
  • Potent SIRT2 inhibition (IC50 7.0 μM).
  • High chemical purity (>99.9%).
  • Solid, white to off-white appearance for easy handling.
  • Suitable for neurodegeneration and SIRT2 biology research.
  • Storage conditions provided to preserve stability and activity.
Catalog No. 50-002-58837 Supplier Medchemexpress LLC Supplier No. HY14723310MG
MIND4-19 is a small-molecule SIRT2 inhibitor (IC50 7.0 μM) used as a research tool in studies of neurodegeneration, including Huntington's disease. The compound is a solid, white to off-white material with formula C22H19N3OS and a molecular weight of 373.47 g·mol⁻¹; it is supplied with high purity and storage recommendations to protect activity.
  • Potent SIRT2 inhibition (IC50 7.0 μM).
  • High chemical purity (>99.9%).
  • Solid, white to off-white appearance for easy handling.
  • Suitable for neurodegeneration and SIRT2 biology research.
  • Storage conditions provided to preserve stability and activity.