Learn More
Glen Research 3'-BHQ-1 CPG, 1 gram
Catalog No. NC2753642
Encompass_Preferred
4'-(2-Nitro-4-toluyldiazo)-2'-methoxy-5'-methyl-azobenzene-4''-(N-ethyl-2-O-(4,4'-dimethoxytrityl))-N-ethyl-2-O-glycolate-CPGF.W.: 554.49Pore Size: 1000Å
Catalog No. NC2753642 Supplier Glen Research Supplier No. 20593110
4'-(2-Nitro-4-toluyldiazo)-2'-methoxy-5'-methyl-azobenzene-4''-(N-ethyl-2-O-(4,4'-dimethoxytrityl))-N-ethyl-2-O-glycolate-CPGF.W.: 554.49Pore Size: 1000Å