Learn More
New England Biolabs, Inc. 3'-Biotin-GTP – 0.5 µmol
SDP
Catalog No. 502024048
Encompass
Supplied in: Ultrapure water as a Triethylammonium salt at pH 7.0 Molecular Formula: C31H50N11O19P3S Molecular Weight: 1005.78 g/mol Extinction Coefficient: 260 = ~11,700 Lmol-1 cm-1
Catalog No. 50-202-4048 Supplier New England Biolabs, Inc. Supplier No. N0760S
Supplied in: Ultrapure water as a Triethylammonium salt at pH 7.0 Molecular Formula: C31H50N11O19P3S Molecular Weight: 1005.78 g/mol Extinction Coefficient: 260 = ~11,700 Lmol-1 cm-1