Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Biphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B24895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24895G 5 g
B2489-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24895G Supplier TCI America Supplier No. B24895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5122-95-2
Molecular Formula C12H11BO2
MDL Number MFCD01318102
InChI Key GOXICVKOZJFRMB-UHFFFAOYSA-N
PubChem CID 2734315

Specifications

Melting Point 208°C
Quantity 5 g
Synonym 3-biphenylboronic acid, biphenyl-3-boronic acid, 1,1'-biphenyl-3-ylboronic acid, biphenyl-3-ylboronic acid, 3-phenylphenyl boranediol, 3-phenylphenyl boronic acid, 3-biphenyl boronic acid, 3-phenylbenzeneboronic acid, 3-biphenylylboronic acid, 1,1'-biphenyl-3-yl boronic acid
SMILES B(C1=CC(=CC=C1)C2=CC=CC=C2)(O)O
IUPAC Name (3-phenylphenyl)boronic acid
Molecular Weight (g/mol) 198.028
Formula Weight 198.03
Physical Form Crystalline Powder
Chemical Name or Material 3-Biphenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.