Learn More
Medchemexpress LLC 3α-bis-(4-fluorophenyl) methoxytropane hydrochloride | 202646-03-5 | 100.0% | 379.87 g/mol | C21H24ClF2NO | 1 ML
SDP
Catalog No. 5000211410
Encompass_Preferred
AHN 1-055 hydrochloride is a dopamine uptake inhibitor that binds with high affinity to the dopamine transporter (DAT). It is provided as a 10 mM solution in DMSO for use in pharmacology and neuroscience research probing DAT function and dopamine reuptake. Key chemical identifiers include CAS 202646-03-5 and molecular weight 379.87 g/mol.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • High potency DAT inhibitor (IC50 = 71 nM).
  • High purity suitable for research use (~99.96%).
  • Compact 1 mL format for small-volume assays.
Catalog No. 50-002-11410 Supplier Medchemexpress LLC Supplier No. HY10131510MM 1ML
AHN 1-055 hydrochloride is a dopamine uptake inhibitor that binds with high affinity to the dopamine transporter (DAT). It is provided as a 10 mM solution in DMSO for use in pharmacology and neuroscience research probing DAT function and dopamine reuptake. Key chemical identifiers include CAS 202646-03-5 and molecular weight 379.87 g/mol.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • High potency DAT inhibitor (IC50 = 71 nM).
  • High purity suitable for research use (~99.96%).
  • Compact 1 mL format for small-volume assays.