Learn More
Medchemexpress LLC 3α-bis-(4-fluorophenyl) methoxytropane hydrochloride | 202646-03-5 | MFCD00671778 | 100.0% | 379.87 g·mol⁻¹ | C21H24ClF2NO | 10 MG
SDP
Catalog No. 5000212423
Encompass_Preferred
AHN 1-055 hydrochloride is a selective dopamine uptake inhibitor supplied as the hydrochloride salt for research use. It binds with high affinity to the dopamine transporter (DAT) and is characterized by an IC50 of 71 nM. The compound is provided as a high-purity solid suitable for biochemical and pharmacological studies.
  • Selective dopamine transporter uptake inhibitor (IC50 71 nM).
  • Provided as the hydrochloride salt in solid form for solution preparation.
  • High purity (99.96%) appropriate for research applications.
  • Available in small-scale quantities suitable for laboratory experiments.
  • Useful for studies of dopamine transporter function and neuropharmacology.
Catalog No. 50-002-12423 Supplier Medchemexpress LLC Supplier No. HY10131510MG
AHN 1-055 hydrochloride is a selective dopamine uptake inhibitor supplied as the hydrochloride salt for research use. It binds with high affinity to the dopamine transporter (DAT) and is characterized by an IC50 of 71 nM. The compound is provided as a high-purity solid suitable for biochemical and pharmacological studies.
  • Selective dopamine transporter uptake inhibitor (IC50 71 nM).
  • Provided as the hydrochloride salt in solid form for solution preparation.
  • High purity (99.96%) appropriate for research applications.
  • Available in small-scale quantities suitable for laboratory experiments.
  • Useful for studies of dopamine transporter function and neuropharmacology.