Learn More
Medchemexpress LLC 3α-bis-(4-fluorophenyl) methoxytropane hydrochloride | 202646-03-5 | MFCD00671778 | 100.0% | 379.87 g/mol | C21H24ClF2NO | 50 MG
SDP
Catalog No. 5000213257
Encompass_Preferred
AHN 1-055 hydrochloride is a dopamine uptake inhibitor with an IC50 of 71 nM that binds with high affinity to the dopamine transporter (DAT). It is supplied as a high-purity solid suitable for in vitro pharmacology and biochemical studies.
  • High affinity inhibitor of the dopamine transporter (IC50 ~71 nM).
  • High purity solid for reliable experimental results (99.96%).
  • White to yellow solid, stable under recommended storage conditions.
  • Available as a solid and as DMSO solutions for flexible use in assays.
  • Suitable for in vitro uptake and receptor binding studies.
Catalog No. 50-002-13257 Supplier Medchemexpress LLC Supplier No. HY10131550MG
AHN 1-055 hydrochloride is a dopamine uptake inhibitor with an IC50 of 71 nM that binds with high affinity to the dopamine transporter (DAT). It is supplied as a high-purity solid suitable for in vitro pharmacology and biochemical studies.
  • High affinity inhibitor of the dopamine transporter (IC50 ~71 nM).
  • High purity solid for reliable experimental results (99.96%).
  • White to yellow solid, stable under recommended storage conditions.
  • Available as a solid and as DMSO solutions for flexible use in assays.
  • Suitable for in vitro uptake and receptor binding studies.