Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Boc-3-azaspiro[5.5]undecane-9-carbaldehyde | 1416176-14-1 | 281.39 g/mol | C16H27NO3
SDP

Catalog No. 5000210003
Encompass_Preferred

3-Boc-3-azaspiro[5.5]undecane-9-carbaldehyde is an aldehyde-functionalized azaspiro scaffold used as a PROTAC linker for assembling bifunctional degraders in targeted protein degradation research.

  • Aldehyde functionality for conjugation reactions.
  • Azaspiro[5.5] scaffold provides conformational constraint.
  • Molecular formula: C16H27NO3.
  • Molecular weight: 281.39 g/mol.
  • Storage: -20°C under nitrogen; in solvent: -80°C for 6 months or -20°C for 1 month (under nitrogen).

Catalog No. 50-002-10003 Supplier Medchemexpress LLC Supplier No. HYW056267100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Boc-3-azaspiro[5.5]undecane-9-carbaldehyde is an aldehyde-functionalized azaspiro scaffold used as a PROTAC linker for assembling bifunctional degraders in targeted protein degradation research.

  • Aldehyde functionality for conjugation reactions.
  • Azaspiro[5.5] scaffold provides conformational constraint.
  • Molecular formula: C16H27NO3.
  • Molecular weight: 281.39 g/mol.
  • Storage: -20°C under nitrogen; in solvent: -80°C for 6 months or -20°C for 1 month (under nitrogen).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.