Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-1,1,1-trifluoro-2-propanol, 96%

Catalog No. AAL1056603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1056603 1 g
AAL1056606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1056603 Supplier Thermo Scientific Chemicals Supplier No. L1056603
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 431-34-5 | C3H4BrF3O | 192.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 431-34-5
Molecular Formula C3H4BrF3O
Molecular Weight (g/mol) 192.96
MDL Number MFCD00040945
InChI Key VBHIIZIQRDVGDH-UHFFFAOYNA-N
PubChem CID 220632
IUPAC Name 3-bromo-1,1,1-trifluoropropan-2-ol
SMILES OC(CBr)C(F)(F)F

Specifications

Density 1.861
Boiling Point 124°C to 126°C
Flash Point 43°C (109°F)
Refractive Index 1.399
Quantity 1 g
UN Number UN1987
Beilstein 1737803
Synonym 3-bromo-1,1,1-trifluoro-2-propanol, pubchem12626, acmc-20an9c, 1,1,1-trifluoro-3-bromo-propan-2-ol, 2-propanol,3-bromo-1,1,1-trifluoro, 3-bromo-1,1,1-trifluoro-propan-2-ol, 2-propanol, 3-bromo-1,1,1-trifluoro, rac-3-bromo-1,1,1-trifluoro-propan-2-ol, 3-bromanyl-1,1,1-tris fluoranyl propan-2-ol
Formula Weight 192.97
Percent Purity 96%
Chemical Name or Material 3-Bromo-1,1,1-trifluoro-2-propanol
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.