Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Bromo-1,4-di(9-carbazolyl)benzene 98.0+%, TCI America™
SDP

Catalog No. B5027200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5027200MG 200 mg
B50271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5027200MG Supplier TCI America Supplier No. B5027200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1537218-76-0
Molecular Formula C30H19BrN2
Molecular Weight (g/mol) 487.4
MDL Number MFCD26406219
InChI Key NEYCIRFFXNWOAH-UHFFFAOYSA-N
Synonym 3-Bromo-9-[4-(9H-carbazol-9-yl)phenyl]-9H-carbazole
PubChem CID 86346300
IUPAC Name 3-bromo-9-(4-carbazol-9-ylphenyl)carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)N5C6=C(C=C(C=C6)Br)C7=CC=CC=C75

Specifications

Melting Point 219°C
Color White-Yellow
Quantity 200 mg
Formula Weight 487.40
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Bromo-1,4-di(9-carbazolyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.