Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-1-(triisopropylsilyl)indole 95.0+%, TCI America™
SDP

Catalog No. B20571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B20571G Supplier TCI America Supplier No. B20571G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 148249-36-9
Molecular Formula C17H26BrNSi
Molecular Weight (g/mol) 352.39
MDL Number MFCD02093441
InChI Key VELAHBNAGHGPNP-UHFFFAOYSA-N
PubChem CID 10991769
IUPAC Name 3-bromo-1-[tris(propan-2-yl)silyl]-1H-indole
SMILES CC(C)[Si](C(C)C)(C(C)C)N1C=C(Br)C2=CC=CC=C12

Specifications

Melting Point 65°C
Color White-Yellow
Quantity 1 g
Formula Weight 352.39
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-1-(triisopropylsilyl)indole
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.