Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Bromo-1-(trimethylsilyl)-1-propyne

Catalog No. 3777751G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-777-51G 1 g
37-777-510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-777-51G Supplier Sigma Aldrich Supplier No. 3777751G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-bromo-1-(trimethylsilyl)-1-propyne is reported as a propargylating agent
  • Selective reaction of 3-bromo-1-(trimethylsilyl)-1-propyne with α-keto ester using indium metal has been studied.3-bromo-1-(trimethylsilyl)-1-propyne may be used in the preparation of tetrahydroisoquinoline-3-carboxylic acid derivatives
  • It may be used as reagent for the alkylation of dianion of α-keto esters at α-carbon
  • It may be employed in the preparation of terminal conjugated enynes and allenic alcohols

Specifications

CAS 38002-45-8
Density 1.17 g/mL (at 25°C (literature))
Boiling Point 44°C to 45°C (2 mmHg)
Molecular Formula C6H11BrSi
Refractive Index n20/D 1.483 (literature)
Linear Formula BrCH2C///CSi(CH3)3
MDL Number MFCD00134460
Quantity 1 g
Synonym 3-(Trimethylsilyl)propargyl bromide; 3-Bromo-1-(trimethylsilyl)-1-propyne
Molecular Weight (g/mol) 191.14
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.