Learn More
Medchemexpress LLC 3-bromo-1H-indazole-7-carbonitrile | 945762-00-5 | MFCD11848409 | 99.5% | 222.04 | C8H4BrN3 | 250 MG
SDP
Catalog No. 5000302450
Encompass_Preferred
nNOS-IN-1 is a small-molecule inhibitor of neuronal nitric oxide synthase (nNOS) used in biochemical and pharmacological research to probe nitric oxide signaling. It is supplied as a solid off-white to light-brown powder with high chemical purity (≈99.5%), molecular formula C8H4BrN3, and molecular weight 222.04. Solubility and storage guidance support both in vitro and in vivo formulation workflows.
  • Selective neuronal nitric oxide synthase inhibitor for research applications.
  • High purity (~99.5%) suitable for biochemical and pharmacological assays.
  • Solid powder appearance simplifies handling and accurate dosing.
  • Soluble in DMSO at high concentrations; compatible with common in vivo vehicles.
  • Stable under recommended storage: powder at -20°C (years) or 4°C (months); solutions at -80°C.
Catalog No. 50-003-02450 Supplier Medchemexpress LLC Supplier No. 250MGHYW022047
nNOS-IN-1 is a small-molecule inhibitor of neuronal nitric oxide synthase (nNOS) used in biochemical and pharmacological research to probe nitric oxide signaling. It is supplied as a solid off-white to light-brown powder with high chemical purity (≈99.5%), molecular formula C8H4BrN3, and molecular weight 222.04. Solubility and storage guidance support both in vitro and in vivo formulation workflows.
  • Selective neuronal nitric oxide synthase inhibitor for research applications.
  • High purity (~99.5%) suitable for biochemical and pharmacological assays.
  • Solid powder appearance simplifies handling and accurate dosing.
  • Soluble in DMSO at high concentrations; compatible with common in vivo vehicles.
  • Stable under recommended storage: powder at -20°C (years) or 4°C (months); solutions at -80°C.