Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2,4-dimethyl-5-phenylthiophene 98.0+%, TCI America™
SDP

Catalog No. B26771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26771G 1 g
B26775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26771G Supplier TCI America Supplier No. B26771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 362513-28-8
Molecular Formula C12H11BrS
Molecular Weight (g/mol) 267.184
InChI Key VJMJMLJTIYJVQY-UHFFFAOYSA-N
PubChem CID 44629772
IUPAC Name 3-bromo-2,4-dimethyl-5-phenylthiophene
SMILES CC1=C(SC(=C1Br)C)C2=CC=CC=C2

Specifications

Quantity 1 g
Formula Weight 267.18
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 3-Bromo-2,4-dimethyl-5-phenylthiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.