Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2,5-diaminopyridine, 99%

Catalog No. AC360660010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC360660010 1 g Glass bottle
AC360660050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC360660010 Supplier Thermo Scientific Chemicals Supplier No. 360660010
Only null left
Add to Cart

CAS: 896160-69-3 | C5H6BrN3 | 188.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 896160-69-3
Molecular Formula C5H6BrN3
Molecular Weight (g/mol) 188.03
MDL Number MFCD03427651
InChI Key YJENCMIONVUOCW-UHFFFAOYSA-N
Synonym 2,5-diamino-3-bromopyridine, 3-bromo-2,5-diaminopyridine, 2,5-diamino-3-bormopyridine, 2,5-pyridinediamine, 3-bromo, pubchem5490, acmc-1c3l9, 3-bromanylpyridine-2,5-diamine, abbypharma ap-31-4166
PubChem CID 2734413
IUPAC Name 3-bromopyridine-2,5-diamine
SMILES NC1=CC(Br)=C(N)N=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 117°C to 122°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 188
Percent Purity 99%
Chemical Name or Material 3-Bromo-2, 5-diaminopyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.