Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2,6-dimethoxybenzoic acid, ≥97%, Thermo Scientific™

Catalog No. BTB07593EB
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB07593EB 25 g
BTB07593DA 1 g
BTB07593EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTB07593EB Supplier Thermo Scientific Chemicals Supplier No. BTB07593EB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73219-89-3
Molecular Formula C9H9BrO4
Molecular Weight (g/mol) 261.071
MDL Number MFCD00052933
InChI Key CUQANLQRQJHIQE-UHFFFAOYSA-N
PubChem CID 2774744
IUPAC Name 3-bromo-2,6-dimethoxybenzoic acid
SMILES COC1=C(C(=C(C=C1)Br)OC)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Quantity 25 g
Synonym maybridge1_001596, acmc-1bfe7, 2,6-dimethoxy-3-bromobenzoic acid, benzoic acid,3-bromo-2,6-dimethoxy, benzoic acid, 3-bromo-2,6-dimethoxy
Formula Weight 261.07
Percent Purity 97%
Chemical Name or Material 3-Bromo-2, 6-dimethoxybenzoic acid
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.