Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-(bromomethyl)propionic acid, 98%

Catalog No. AAA1963022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1963022 100 g
AAA1963006 5 g
AAA1963014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1963022 Supplier Thermo Scientific Chemicals Supplier No. A1963022
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 41459-42-1 | C4H5Br2O2 | 244.89 g/mol

3-Bromo-2-(bromomethyl)propionic acid acts as an organic building block for the preparation of beta-substituted acrylates. It is used in the preparation of t-butyl 2-(phenylthiomethyl) propenoate, t-butyl 3-(phenylthio)-2-(phenylthiomethyl)propenoate and 3-(phenylthio)-2-(phenyl-sulfinylmethyl)propenoate. Further, it plays an important role in the synthesis of beta-lactams by cyclization of the corresponding amide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2-(bromomethyl)propionic acid acts as an organic building block for the preparation of beta-substituted acrylates. It is used in the preparation of t-butyl 2-(phenylthiomethyl) propenoate, t-butyl 3-(phenylthio)-2-(phenylthiomethyl)propenoate and 3-(phenylthio)-2-(phenyl-sulfinylmethyl)propenoate. Further, it plays an important role in the synthesis of beta-lactams by cyclization of the corresponding amide.

Solubility
Soluble in acetic acid.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41459-42-1
Molecular Formula C4H5Br2O2
Molecular Weight (g/mol) 244.89
MDL Number MFCD00010643
InChI Key QQZJWQCLWOQDQV-UHFFFAOYSA-M
Synonym 3-bromo-2-bromomethyl propionic acid, 3-bromo-2-bromomethyl propanoic acid, propanoic acid, 3-bromo-2-bromomethyl, .beta.,.beta.-dibromoisobutyric acid, 3-bromo-2-bromomethyl propionicacid, bis bromomethyl acetic acid, zlchem 161, acmc-209jkh, beta,beta'-dibromoisobutyric acid, 2-bromomethyl-3-bromopropanoic acid
PubChem CID 318915
IUPAC Name 3-bromo-2-(bromomethyl)propanoic acid
SMILES [O-]C(=O)C(CBr)CBr

Specifications

Melting Point 98°C to 101°C
Quantity 100 g
UN Number UN3261
Solubility Information Soluble in acetic acid.
Formula Weight 245.91
Percent Purity 98%
Chemical Name or Material 3-Bromo-2-(bromomethyl)propionic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.