Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-fluorobenzaldehyde, 97%

Catalog No. AAH6465022
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6465022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6465022 Supplier Thermo Scientific Chemicals Supplier No. H6465022

missing translation for 'validMainframeMsg'

CAS: 149947-15-9 | C7H4BrFO | 203.01 g/mol

It can be used as pharmaceutical raw material and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149947-15-9
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD09027089
InChI Key OUAZPCKRSSEQKB-UHFFFAOYSA-N
Synonym 3-bromo-2-fluorobenzenecarbaldehyde, 3-bromo-2-fluorobenzadehyde, 2-fluoro-3-bromobenzaldehyde, benzaldehyde,3-bromo-2-fluoro, benzaldehyde, 3-bromo-2-fluoro, 3-bromofluorobenzaldehyde, intermediates-zcf02145, acmc-209d3i, 3-bromo-2-fluoro-benzaldehyde, 3-bromanyl-2-fluoranyl-benzaldehyde
PubChem CID 22019361
IUPAC Name 3-bromo-2-fluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)Br)F)C=O

Specifications

Melting Point 43°C to 46°C
Color White
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 13904499
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 203.01
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Bromo-2-fluorobenzaldehyde
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.