Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-fluorobenzamide, 96%, Thermo Scientific™

Catalog No. AAH62619MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62619MD 250 mg
AAH6261903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62619MD Supplier Thermo Scientific Chemicals Supplier No. H62619MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871353-25-2
Molecular Formula C7H5BrFNO
Molecular Weight (g/mol) 218.03
MDL Number MFCD12913463
InChI Key UWTBFBQQCCZYBF-UHFFFAOYSA-N
Synonym benzamide, 3-bromo-2-fluoro
PubChem CID 44203102
IUPAC Name 3-bromo-2-fluorobenzamide
SMILES NC(=O)C1=C(F)C(Br)=CC=C1

Specifications

Quantity 250 mg
Formula Weight 218.02
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Bromo-2-fluorobenzamide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.