Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B51371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51371G 1 g
B5137-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51371G Supplier TCI America Supplier No. B51371G
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 3-Bromo-2-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 352535-97-8
Melting Point 227°C
Color White-Yellow
Molecular Formula C6H5BBrFO2
Quantity 1 g
MDL Number MFCD05664228
Synonym 3-bromo-2-fluorophenyl boronic acid, 2-fluoro-3-bromophenylboronic acid, boronic acid,b-3-bromo-2-fluorophenyl, boronic acid, b-3-bromo-2-fluorophenyl, 3-bromo-2-fluorophenylboronicacid, acmc-1aijv, 3-bromo-2-fluoro-phenylboronic acid, 3-bromo-2-fluorobenzeneboronic acid, 3-bromanyl-2-fluoranyl-phenyl boronic acid, contains varying amounts of anhydride
InChI Key QCLOSORNRRMFIA-UHFFFAOYSA-N
SMILES B(C1=C(C(=CC=C1)Br)F)(O)O
IUPAC Name (3-bromo-2-fluorophenyl)boronic acid
Molecular Weight (g/mol) 218.816
PubChem CID 16217170
Formula Weight 218.82
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.