Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-fluoropyridine, 95%, Thermo Scientific™

Catalog No. AAH2720803
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2720803 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2720803 Supplier Thermo Scientific Chemicals Supplier No. H2720803

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 36178-05-9
Molecular Formula C5H3BrFN
Quantity 1g
MDL Number MFCD04112496
UN Number UN2810
Synonym 2-fluoro-3-bromopyridine, 3-bromo-2-fluoro-pyridine, pyridine, 3-bromo-2-fluoro, bromofluoropyridine, pubchem3008, acmc-1ct2w, pyridine,3-bromo-2-fluoro, ksc576a4j, 3-bromo-2-fluoro pyridine
InChI Key KESUTBOSNOHAMK-UHFFFAOYSA-N
SMILES C1=CC(=C(N=C1)F)Br
IUPAC Name 3-bromo-2-fluoropyridine
Molecular Weight (g/mol) 175.988
PubChem CID 2762778
Formula Weight 175.99
Percent Purity 95%
Chemical Name or Material 3-Bromo-2-fluoropyridine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H311-H227-H302-H332-H315-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.