Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-fluoropyridine 98.0+%, TCI America™
SDP

Catalog No. B37211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37211G 1 g
B37215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37211G Supplier TCI America Supplier No. B37211G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36178-05-9
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.988
MDL Number MFCD04112496
InChI Key KESUTBOSNOHAMK-UHFFFAOYSA-N
PubChem CID 2762778
IUPAC Name 3-bromo-2-fluoropyridine
SMILES C1=CC(=C(N=C1)F)Br

Specifications

Color Yellow
Boiling Point 76°C
Quantity 1 g
Synonym 2-fluoro-3-bromopyridine, 3-bromo-2-fluoro-pyridine, pyridine, 3-bromo-2-fluoro, bromofluoropyridine, pubchem3008, acmc-1ct2w, pyridine,3-bromo-2-fluoro, ksc576a4j, 3-bromo-2-fluoro pyridine
Formula Weight 175.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-fluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.