Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-furoic acid, 97%, Thermo Scientific™

Catalog No. CC67201DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC67201DA 1 g Amber Glass Bottle
CC67201CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC67201DA Supplier Thermo Scientific Chemicals Supplier No. CC67201DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14903-90-3
Molecular Formula C5H3BrO3
Molecular Weight (g/mol) 190.98
MDL Number MFCD00092761
InChI Key UZBGSJZFBUOJNE-UHFFFAOYSA-N
PubChem CID 605479
IUPAC Name 3-bromofuran-2-carboxylic acid
SMILES C1=COC(=C1Br)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-bromo-2-furoic acid, furancarboxylic acid, bromo, 3-bromo-2-furancarboxylic acid, 3-bromo-furan-2-carboxylic acid, 3-bromofuran-2-carboxylicacid, bromofuroic acid, 3-bomo-2-furoic acid, 2-furancarboxylic acid,3-bromo, 3-bromanylfuran-2-carboxylic acid, 2-furancarboxylic acid, 3-bromo
Formula Weight 190.98
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-furoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.