Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-hydroxy-5-nitropyridine, 98%, Thermo Scientific™

Catalog No. AAH6673906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6673906 5 g
AAH6673914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6673906 Supplier Thermo Scientific Chemicals Supplier No. H6673906

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15862-33-6
Molecular Formula C5H3BrN2O3
Molecular Weight (g/mol) 218.994
MDL Number MFCD03840431
InChI Key KWSOHRDMTWDAOI-UHFFFAOYSA-N
Synonym 3-bromo-2-hydroxy-5-nitropyridine, 3-bromo-5-nitropyridin-2-ol, 2-hydroxy-3-bromo-5-nitropyridine, 3-bromo-5-nitro-2 1h-pyridinone, 3-bromo-5-nitro-2-pyridinol, 2 1h-pyridinone, 3-bromo-5-nitro, 3-bromo-5-nitro-2-hydroxypyridine, pubchem6708, acmc-209dic, ksc684c2p
PubChem CID 613279
IUPAC Name 3-bromo-5-nitro-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1[N+](=O)[O-])Br

Specifications

Melting Point 213°C to 218°C
Quantity 5 g
Beilstein 135472
Formula Weight 218.99
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Bromo-2-hydroxy-5-nitropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.