Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Bromo-2'-methoxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. B5498200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5498200MG 200 mg
B54981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5498200MG Supplier TCI America Supplier No. B5498200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 267651-23-0
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD22573042
InChI Key IBHCVANJIBGASI-UHFFFAOYSA-N
Synonym 1-(3-Bromo-2-methoxyphenyl)ethanone
PubChem CID 59544891
IUPAC Name 1-(3-bromo-2-methoxyphenyl)ethanone
SMILES CC(=O)C1=C(C(=CC=C1)Br)OC

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 229.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3′-Bromo-2′-methoxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.