Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 3-Bromo-2-methoxypyridine, 98%, [13472-59-8], B12933 188.022 C6H6BrNO

Catalog No. 501374699
Encompass_Preferred

Compound: 3-Bromo-2-methoxypyridine

CAS#: 13472-59-8

MDL#: not listed

Background: A brominated, methoxylated pyridine used as a common building block in pharmaceutical research. The bromine substituent offers a reactive site for cross-coupling, a standard transformation for elaborating pyridine scaffolds.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-137-4699 Supplier Frontier Specialty Chemicals Supplier No. B1293325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Bromo-2-methoxypyridine

CAS#: 13472-59-8

MDL#: not listed

Background: A brominated, methoxylated pyridine used as a common building block in pharmaceutical research. The bromine substituent offers a reactive site for cross-coupling, a standard transformation for elaborating pyridine scaffolds.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.