Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Bromo-2-methylpropene

Catalog No. 3458305ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-583-05ML 5 mL
34-583-025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 34-583-05ML Supplier Sigma Aldrich Supplier No. 3458305ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Rotational isomerism in 3-bromo-2-methylpropene has been studied by IR spectroscopy.3-bromo-2-methylpropene may be used:
  • In the synthesis of alkenyl imines
  • In the synthesis of 2-methylpropenyl (methallyl) complex, cp*os(++3-allyl)br2
  • In a study of chiral phase transfer alkylation leading to (s)-α-alkylcysteines

Specifications

CAS 1458-98-6
Density 1.339 g/mL (at 25°C (literature))
Boiling Point 94°C to 95°C (lit.)
Molecular Formula C4H7Br
Refractive Index n20/D 1.472 (lit.)
Linear Formula H2C=C(CH3)CH2Br
MDL Number MFCD00134155
Quantity 5 mL
Synonym Methallyl bromide; Methylallyl bromide
Molecular Weight (g/mol) 135
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.