Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 3-Bromo-2-methylpropene, 1458-98-6, MFCD00134155, 5g

Catalog No. 500127362
Encompass_Preferred

Molecular Formula C4H7Br, Purity 95%, Molecular Weight 135.01, Boiling Point 94-95

Catalog No. 50-012-7362 Supplier Matrix Scientific Supplier No. 0182535G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C4H7Br, Purity 95%, Molecular Weight 135.01, Boiling Point 94-95

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.