Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Bromo-2-methylpyridine | 38749-79-0 | 191224 | 172.02 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW002494

Encompass_Preferred

3-Bromo-2-methylpyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. This product is for research use only and not sold to patients.

  • Molecular weight: 172.02
  • Formula: C6H6BrN
  • Appearance: liquid
  • Color: colorless to light yellow
  • Density: 1.495 g/cm³

Catalog No. 50-004-49466


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.