Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. B29621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29621G 1 g
B29625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29621G Supplier TCI America Supplier No. B29621G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38749-79-0
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00191224
InChI Key AIPWPTPHMIYYOX-UHFFFAOYSA-N
Synonym 3-bromo-2-picoline, 2-methyl-3-bromopyridine, 3-bromo-2-methyl-pyridine, pyridine, 3-bromo-2-methyl, 2-methyl-3-bromo pyridine, pubchem1220, 3-brom-2-methylpyridin, acmc-209j1t, 3-bromo-2-methyl pyridine, ksc497m1l
PubChem CID 1201003
IUPAC Name 3-bromo-2-methylpyridine
SMILES CC1=C(Br)C=CC=N1

Specifications

Color Yellow
Boiling Point 76°C
Quantity 1 g
Formula Weight 172.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.