Learn More
Medchemexpress LLC 3-bromo-4-chlorobenzoic acid | 42860-10-6 | MFCD00079706 | 100.0% | 235.46 g·mol⁻¹ | C7H4BrClO2 | 50 G
SDP
Catalog No. 5000301070
Encompass_Preferred
3-Bromo-4-chlorobenzoic acid is a brominated and chlorinated benzoic acid used as an intermediate in organic and pharmaceutical synthesis. It is reported as Dapagliflozin Impurity 50 and is supplied for research, method development, and impurity profiling, with high listed purity and multiple packaging options.
  • High purity suitable for synthetic applications.
  • Useful intermediate for medicinal chemistry and impurity studies.
  • Defined molecular formula and molecular weight for analytical work.
  • Available in a range of packaging sizes for scale flexibility.
  • Suitable as an analytical reference standard in method development.
Catalog No. 50-003-01070 Supplier Medchemexpress LLC Supplier No. HYW01869750G
Add to Cart
Add to Cart
3-Bromo-4-chlorobenzoic acid is a brominated and chlorinated benzoic acid used as an intermediate in organic and pharmaceutical synthesis. It is reported as Dapagliflozin Impurity 50 and is supplied for research, method development, and impurity profiling, with high listed purity and multiple packaging options.
  • High purity suitable for synthetic applications.
  • Useful intermediate for medicinal chemistry and impurity studies.
  • Defined molecular formula and molecular weight for analytical work.
  • Available in a range of packaging sizes for scale flexibility.
  • Suitable as an analytical reference standard in method development.