Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-fluorobenzaldehyde, 99%

Catalog No. AC293930250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC293930250 25 g
AC293930050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC293930250 Supplier Thermo Scientific Chemicals Supplier No. 293930250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 77771-02-9 | C7H4BrFO | 203.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77771-02-9
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD00042186
InChI Key FAHZIKXYYRGSHF-UHFFFAOYSA-N
Synonym 4-fluoro-3-bromobenzaldehyde, benzaldehyde, 3-bromo-4-fluoro, 3-bromo-4-fluoro-benzaldehyde, 3-bromo-4-fluoro benzaldehyde, pubchem2878, acmc-209pbm, 3-bromo4fluorobenzaldehyde, 3-bromo-4fluorobenzaldehyde, 3-bromo4-fluorobenzaldehyde, 3bromo-4-fluorobenzaldehyde
PubChem CID 173604
IUPAC Name 3-bromo-4-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)Br)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 28°C to 33°C
Color White to Yellow
Boiling Point 138°C to 139°C (2.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Refractive Index 1.574
Linear Formula BrC6H3(F)CHO
Quantity 25 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 203.01
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material 3-Bromo-4-fluorobenzaldehyde, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 278-764-1
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.