Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-fluorobenzylamine hydrochloride, 98%, Thermo Scientific™

Catalog No. AC290510100
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC290510100 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC290510100 Supplier Thermo Scientific Chemicals Supplier No. 290510100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 77771-03-0
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C7H6BrFO
Linear Formula BrC6H3(F)CH2NH2·HCl
Quantity 10g
MDL Number MFCD00143094
Synonym 3-bromo-4-fluorobenzyl alcohol, 3-bromo-4-fluorophenyl methanol, 3-bromo-4-fluorobenzylalcohol, 3-bromo-4-fluoro-phenyl-methanol, 3-bromo-4-fluorophenyl methan-1-ol, pubchem4884, 4-fluoro-3-bromobenzyl alcohol, 3-bromo-4-fluoro-benzyl alcohol, 4-fluoro-3-bromo-benzyl alcohol, 3-bromo-4-fluoro benzyl alcohol
InChI Key HNVVROFWONXXGO-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1CO)Br)F
IUPAC Name (3-bromo-4-fluorophenyl)methanol
Molecular Weight (g/mol) 205.026
PubChem CID 2773351
Formula Weight 240.5
Percent Purity ≥97.5%
Chemical Name or Material 3-Bromo-4-fluorobenzylamine hydrochloride, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.