Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-fluorophenylacetic acid, 96%, Thermo Scientific™

Catalog No. AAH6362203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6362203 1 g
AAH6362206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6362203 Supplier Thermo Scientific Chemicals Supplier No. H6362203

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Bromo-4-fluorophenylacetic Acid is used as a reagent in the preparation of 4-oxo-1,4-dihydripyridine-3-carboxamide derivatives as inhibitors of Axl tyrosine kinase and anticancer agents.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 194019-11-9
Molecular Formula C8H6BrFO2
Molecular Weight (g/mol) 233.04
MDL Number MFCD04038373
InChI Key XXFGIJYSXNXNAU-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorophenylacetic acid, 2-3-bromo-4-fluorophenyl acetic acid, 3-bromo-4-fluorophenylaceticacid, 3-bromo-4-fluorophenyl acetic acid, benzeneacetic acid, 3-bromo-4-fluoro, 3-bromo-4-fluoro-phenyl-acetic acid, 2-3-bromo-4-fluoro-phenyl acetic acid, acmc-209exn, ksc497i4b, 61150-59-2 3-bromo-4-fluorophenylacetic acid
PubChem CID 3416339
IUPAC Name 2-(3-bromo-4-fluorophenyl)acetic acid
SMILES OC(=O)CC1=CC(Br)=C(F)C=C1

Specifications

Melting Point 65°C to 69°C (decomposition)
Quantity 1 g
Beilstein 2446892
Solubility Information Slightly soluble in water.
Formula Weight 233.04
Percent Purity 96%
Chemical Name or Material 3-Bromo-4-fluorophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.