Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-methoxybenzyl Cyanide 98.0+%, TCI America™
SDP

Catalog No. B14555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14555G 5 g
B145525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14555G Supplier TCI America Supplier No. B14555G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 772-59-8
Molecular Formula C9H8BrNO
Molecular Weight (g/mol) 226.07
MDL Number MFCD00016391
InChI Key OBJKHHRZMIIEOK-UHFFFAOYSA-N
Synonym 3-bromo-4-methoxyphenylacetonitrile, 3-bromo-4-methoxybenzyl cyanide, 2-3-bromo-4-methoxyphenyl acetonitrile, benzeneacetonitrile, 3-bromo-4-methoxy, 2-3-bromo-4-methoxyphenyl ethanenitrile, acmc-209p8t, benzeneacetonitrile,3-bromo-4-methoxy, 3-bromo-4-methoxy-phenyl-acetonitrile, 2-3-bromanyl-4-methoxy-phenyl ethanenitrile
PubChem CID 522655
IUPAC Name 2-(3-bromo-4-methoxyphenyl)acetonitrile
SMILES COC1=C(Br)C=C(CC#N)C=C1

Specifications

Melting Point 56°C
Color Yellow
Boiling Point 178°C
Quantity 5 g
UN Number 2811
Formula Weight 226.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-methoxybenzyl Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.