Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-methoxyphenylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. B46771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46771G 1 g
B46775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46771G Supplier TCI America Supplier No. B46771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 774-81-2
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.072
MDL Number MFCD00017538
InChI Key POTVGQUUEQTPNA-UHFFFAOYSA-N
PubChem CID 136615
IUPAC Name 2-(3-bromo-4-methoxyphenyl)acetic acid
SMILES COC1=C(C=C(C=C1)CC(=O)O)Br

Specifications

Melting Point 117°C
Color White-Yellow
Quantity 1 g
UN Number 3077
Formula Weight 245.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-methoxyphenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.