Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-5-chlorophenol, ≥97%

Click to view available options
Quantity:
1g
5g
25g
56962-04-0
C6H4BrClO
207.451
MFCD07780676
GMGWXLPFRHYWAS-UHFFFAOYSA-N
phenol, 3-bromo-5-chloro,3-bromo-5-chloro-phenol,3-bromo-5-chlorophenol, 97+%,pubchem17565,acmc-209lv4,ksc268s7h
22630180
3-bromo-5-chlorophenol
C1=C(C=C(C=C1Cl)Br)O
56962-04-0
C6H4BrClO
207.451
MFCD07780676
GMGWXLPFRHYWAS-UHFFFAOYSA-N
phenol, 3-bromo-5-chloro,3-bromo-5-chloro-phenol,3-bromo-5-chlorophenol, 97+%,pubchem17565,acmc-209lv4,ksc268s7h
22630180
3-bromo-5-chlorophenol
C1=C(C=C(C=C1Cl)Br)O
Quantity 5g
Melting Point 66°C to 70°C
Beilstein 3234780
Formula Weight 207.45
Physical Form Solid
Percent Purity ≥97%
Chemical Name or Material 3-Bromo-5-chlorophenol

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

storageNote1 : 2° to 8°C