Learn More
Medchemexpress LLC 3-Bromo-5-chloropyridine | 73583-39-8 | 192.44 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00316510G
Encompass_Preferred
3-Bromo-5-chloropyridine is a widely used heterocyclic compound. It can be utilized in the synthesis of various compounds and serves as an intermediate in the production of crop protectants for the agricultural chemicals industry. This compound also participates in cross-coupling reactions. It appears as an off-white to light brown solid.
  • Can be used to synthesize various compounds.
  • Serves as an intermediate in crop protectant production.
  • Participates in cross-coupling reactions.
  • Appears as an off-white to light brown solid.
  • For research use only.
Catalog No. 50-004-48693