Learn More
eMolecules​ 1208075-19-7 | 3-Bromo-5-ethyltoluene | Oakwood Chemical | MFCD12547777 | 199.091 | C9H11Br | 0.000 | CCc1cc(C)cc(Br)c1 | 1g | 537689685
Catalog No. 502142284 Shop All eMolecules Inc. Products
Encompass_Preferred
3-Bromo-5-ethyltoluene | Oakwood Chemical | 1208075-19-7 | MFCD12547777 | 199.091 | C9H11Br | 0.000 | CCc1cc(C)cc(Br)c1 | 1g | 537689685
Catalog No. 50-214-2284 Supplier eMolecules​ Supplier No. 537689685
3-Bromo-5-ethyltoluene | Oakwood Chemical | 1208075-19-7 | MFCD12547777 | 199.091 | C9H11Br | 0.000 | CCc1cc(C)cc(Br)c1 | 1g | 537689685