Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-fluorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B37825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37825G 5 g
B37821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37825G Supplier TCI America Supplier No. B37825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 176548-70-2
Molecular Formula C7H3BrFO2
Molecular Weight (g/mol) 218.00
MDL Number MFCD01569540
InChI Key KLSLJMGWUPAQGZ-UHFFFAOYSA-M
Synonym 5-bromo-3-fluorobenzoic acid, 3-fluoro-5-bromobenzoic acid, 3-bromo-5-fluorobenzoicacid, 5-fluoro-3-bromobenzoic acid, 3-bromo-5-fluoro-benzoic acid, 3-bromo-5-fluoro benzoic acid, buttpark 24\01-97, benzoic acid, 3-bromo-5-fluoro, pubchem2154, acmc-1c8tm
PubChem CID 2773339
IUPAC Name 3-bromo-5-fluorobenzoate
SMILES [O-]C(=O)C1=CC(F)=CC(Br)=C1

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 5 g
Formula Weight 219.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-fluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.