Learn More
Matrix Scientific 3-Bromo-5-fluoropyridine, 407-20-5, MFCD04112555, 5g
Supplier: Matrix Scientific 0183505G
Molecular Formula C5H3BrFN, Purity 97%, Molecular Weight 175.99, Melting Point 24-28°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.