Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-hydroxypyridine 98.0+%, TCI America™
SDP

Catalog No. B39695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39695G 5 g
B39691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39695G Supplier TCI America Supplier No. B39695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 74115-13-2
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 174.00
MDL Number MFCD00661305
InChI Key VNYBIBSZZDAEOK-UHFFFAOYSA-N
Synonym 3-bromo-5-hydroxypyridine, 5-bromo-3-pyridinol, 5-bromo-3-hydroxypyridine, 5-bromo-pyridin-3-ol, 3-pyridinol, 5-bromo, 3-bromo-5-pyridinol, 3-hydroxy-5-bromopyridine, 5-bromo-3-hydroxy-pyridine, pyridine, 3-ol-5-bromo, 5-bromopyrid-3-ol
PubChem CID 599529
IUPAC Name 5-bromopyridin-3-ol
SMILES OC1=CN=CC(Br)=C1

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 5 g
Formula Weight 174.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-hydroxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.